3-Methylquinoline 95%

Leverantör: Apollo Scientific

OR911987-25G OR911987-5G
APOSOR911987-25GEA 1400 SEK
APOSOR911987-25G APOSOR911987-5G
3-Methylquinoline 95%
3-Methylquinoline
Formel: C₁₀H₉N
MW: 143,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00014661
CAS nummer: 612-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR