2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)ethanol 95%
Leverantör: Apollo Scientific
OR911975-1G OR911975-250MG
APOSOR911975-1GEA 2900 SEK
APOSOR911975-1G APOSOR911975-250MG
2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)ethanol 95%
2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)ethanol
Formel: C₈H₁₄N₂O
MW: 154,21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08060042
CAS nummer: 1007462-48-7