Methyl pyrazolo[1,5-a]pyridine-3-carboxylate 95%
Leverantör: Apollo Scientific
Synonyms: Pyrazolo[1,5-a]pyridine-3-carboxylic acid methyl ester
OR911837-1G OR911837-250MG
APOSOR911837-1GEA 5125 SEK
APOSOR911837-1G APOSOR911837-250MG
Methyl pyrazolo[1,5-a]pyridine-3-carboxylate 95%
Methyl pyrazolo[1,5-a]pyridine-3-carboxylate
Formel: C₉H₈N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD09752651
CAS nummer: 63237-84-3