2,3-Dimethylphenylacetic acid 98%
Leverantör: Apollo Scientific
Synonyms: 2,3-Dimethylbenzeneacetic acid
OR911756-1G OR911756-25G OR911756-5G
APOSOR911756-1GEA 515 SEK
APOSOR911756-1G APOSOR911756-25G APOSOR911756-5G
2,3-Dimethylphenylacetic acid 98%
2,3-Dimethylphenylacetic acid
Formel: C₁₀H₁₂O₂
MW: 164.204 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03844736
CAS nummer: 30981-98-7