3,3-Pentamethylene glutarimide 98%
Leverantör: Apollo Scientific
Synonyms: Gabapentin Impurity 3
OR911683-100G OR911683-25G OR911683-5G
APOSOR911683-100GEA 6275 SEK
APOSOR911683-100G APOSOR911683-25G APOSOR911683-5G
3,3-Pentamethylene glutarimide 98%
3,3-Pentamethylene glutarimide
Formel: C₁₀H₁₅NO₂
MW: 181,235 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00023872
CAS nummer: 1130-32-1