Ethyl-5-chloro-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Leverantör: Apollo Scientific
Varning
OR9116-100MG OR9116-250MG
APOSOR9116-100MGEA 1525 SEK
APOSOR9116-100MG APOSOR9116-250MG
Ethyl-5-chloro-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl-5-chloro-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₆H₁₃ClN₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD07366503
CAS nummer: 959577-68-5