3-Methyl-2-oxovaleric acid 95%
Leverantör: Apollo Scientific
Synonyms: 3-Methyl-2-oxopentanoic acid
OR911420-1G OR911420-5G
APOSOR911420-1GEA 2425 SEK
APOSOR911420-1G APOSOR911420-5G
3-Methyl-2-oxovaleric acid 95%
3-Methyl-2-oxovaleric acid
Formel: C₆H₁₀O₃
MW: 130,14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00002582
CAS nummer: 1460-34-0