6-Nitro-o-cresol 98%
Leverantör: Apollo Scientific
Synonyms: 2-Methyl-6-nitrophenol
OR911362-100G OR911362-10G OR911362-25G
APOSOR911362-100GEA 10025 SEK
APOSOR911362-100G APOSOR911362-10G APOSOR911362-25G
6-Nitro-o-cresol 98%
6-Nitro-o-cresol
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00031116
CAS nummer: 13073-29-5