2-Phenoxynicotinonitrile 96%
Leverantör: Apollo Scientific
Synonyms:
3-Cyano-2-phenoxypyridine
OR911174-25G
OR911174-5G
APOSOR911174-25GEA
5150
SEK
APOSOR911174-25G
APOSOR911174-5G
2-Phenoxynicotinonitrile 96%
2-Phenoxynicotinonitrile
Formel:
C₁₂H₈N₂O MW: 196,21 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00015473 CAS nummer: 14178-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...