4-Ethyl-1,3-thiazol-2-amine hydrochloride 97%
Leverantör: Apollo Scientific
Synonyms: 2-Amino-4-ethylthiazole
OR910233-1G OR910233-250MG
APOSOR910233-1GEA 4375 SEK
APOSOR910233-1G APOSOR910233-250MG
4-Ethyl-1,3-thiazol-2-amine hydrochloride 97%
4-Ethyl-1,3-thiazol-2-amine hydrochloride
Formel: C₅H₉ClN₂S
MW: 164,66 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD17976987
CAS nummer: 34631-53-3