Benzo[d]isothiazol-3-amine 95%

Leverantör: Apollo Scientific

OR910077-1G OR910077-250MG
APOSOR910077-1GEA 2250 SEK
APOSOR910077-1G APOSOR910077-250MG
Benzo[d]isothiazol-3-amine 95%
Benzo[d]isothiazol-3-amine
Formel: C₇H₆N₂S
MW: 150,2009 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00726295
CAS nummer: 23031-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR