Cyclopentylacetonitrile 98%

Leverantör: Apollo Scientific

OR909764-10G OR909764-1G OR909764-25G
APOSOR909764-10GEA 5300 SEK
APOSOR909764-10G APOSOR909764-1G APOSOR909764-25G
Cyclopentylacetonitrile 98%
Cyclopentylacetonitrile
Formel: C₇H₁₁N
Storage Temperature: Omgivande
MDL Number: MFCD00058671
CAS nummer: 5732-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR