7-Methoxy-2-tetralone 96%

Leverantör: Apollo Scientific
Varning

Synonyms: 7-Methoxy-3,4-dihydronaphthalen-2(1H)-one, 3,4-Dihydro-7-methoxynaphthalen-2(1H)-one

OR9096-25G OR9096-5G OR9096-1G
APOSOR9096-25GEA 4400 SEK
APOSOR9096-25G APOSOR9096-5G APOSOR9096-1G
7-Methoxy-2-tetralone 96%
7-Methoxy-2-tetralone
Formel: C₁₁H₁₂O₂
MW: 176,22 g/mol
Kokpunkt: 124…126 °C/1,5mmHg
Densitet: 1,13 g/cm³ (25 °C)
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00001730
CAS nummer: 4133-34-0
EINECS: 223-954-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR