4,5,6,7-Tetrahydro-1H-indazole 98%
Leverantör: Apollo Scientific
OR909279-100G OR909279-25G OR909279-5G
APOSOR909279-100GEA 10175 SEK
APOSOR909279-100G APOSOR909279-25G APOSOR909279-5G
4,5,6,7-Tetrahydro-1H-indazole 98%
4,5,6,7-Tetrahydro-1H-indazole
Formel: C₇H₁₀N₂
MW: 122,17 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00014565
CAS nummer: 2305-79-5