1-(4-Chlorophenyl)cyclopropanecarboxylic acid 97%
Leverantör: Apollo Scientific
Synonyms: 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid
OR909222-1G OR909222-25G OR909222-5G
APOSOR909222-1GEA 975 SEK
APOSOR909222-1G APOSOR909222-25G APOSOR909222-5G
1-(4-Chlorophenyl)cyclopropanecarboxylic acid 97%
1-(4-Chlorophenyl)cyclopropanecarboxylic acid
Formel: C₁₀H₉ClO₂
MW: 196,64 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00001289
CAS nummer: 72934-37-3