5-Chloro-N,2-dimethylaniline hydrochloride 98%
Leverantör: Apollo Scientific
Synonyms: N-Methyl 5-chloro-2-methylaniline hydrochloride
OR909208-1G OR909208-5G
APOSOR909208-1GEA 2575 SEK
APOSOR909208-1G APOSOR909208-5G
5-Chloro-N,2-dimethylaniline hydrochloride 98%
5-Chloro-N,2-dimethylaniline hydrochloride
Formel: C₈H₁₁Cl₂N
Storage Temperature: Omgivande
MDL Number: MFCD12756441
CAS nummer: 1187386-18-0