3-(2-Pyridyl)propionic acid 98%
Leverantör: Apollo Scientific
Synonyms: 3-(2-Pyridyl)propionic Acid
OR909092-1G OR909092-5G
APOSOR909092-1GEA 975 SEK
APOSOR909092-1G APOSOR909092-5G
3-(2-Pyridyl)propionic acid 98%
3-(2-Pyridyl)propionic acid
Formel: C₈H₉NO₂
MW: 151.16 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00995159
CAS nummer: 15197-75-8