3-Nitrobiphenyl 98%

Leverantör: Apollo Scientific

Synonyms: 3-Nitro-1,1'-biphenyl

OR908979-100G OR908979-10G OR908979-25G
APOSOR908979-100GEA 10350 SEK
APOSOR908979-100G APOSOR908979-10G APOSOR908979-25G
3-Nitrobiphenyl 98%
3-Nitrobiphenyl
Formel: C₁₂H₉NO₂
MW: 199,21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00007245
CAS nummer: 2113-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR