4-Bromobenzothiazole 98%

Leverantör: Apollo Scientific
Varning

OR908977-1G OR908977-250MG OR908977-5G
APOSOR908977-1GEA 1300 SEK
APOSOR908977-1G APOSOR908977-250MG APOSOR908977-5G
4-Bromobenzothiazole 98%
4-Bromobenzothiazole
Formel: C₇H₄BrNS
MW: 214,0800 g/mol
Kokpunkt: 90 °C
Storage Temperature: Kylskåp
MDL Number: MFCD11858590
CAS nummer: 767-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR