Ethyl-4-hydroxycyclohexanecarboxylate 98%
Leverantör: Apollo Scientific
Synonyms: Ethyl 4-hydroxycyclohexanecarboxylate
OR908004-100G OR908004-25G OR908004-5G
APOSOR908004-100GEA 5675 SEK
APOSOR908004-100G APOSOR908004-25G APOSOR908004-5G
Ethyl-4-hydroxycyclohexanecarboxylate 98%
Ethyl-4-hydroxycyclohexanecarboxylate
Formel: C₉H₁₆O₃
MW: 172,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00192156
CAS nummer: 17159-80-7