4-Cyanotetrahydro-4H-pyran 97%

Leverantör: Apollo Scientific

Synonyms: Oxane-4-carbonitrile

OR907988-10G OR907988-25G OR907988-5G
APOSOR907988-10GEA 740 SEK
APOSOR907988-10G APOSOR907988-25G APOSOR907988-5G
4-Cyanotetrahydro-4H-pyran 97%
4-Cyanotetrahydro-4H-pyran
Formel: C₆H₉NO
MW: 111,1400 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08690357
CAS nummer: 4295-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR