2-Methyl-N-tert-butylbenzamide 98%
Leverantör: Apollo Scientific
Synonyms:
N-t-butyl-2-methylbenzamide
OR907676-100G
OR907676-25G
OR907676-5G
APOSOR907676-100GEA
7450
SEK
APOSOR907676-100G
APOSOR907676-25G
APOSOR907676-5G
2-Methyl-N-tert-butylbenzamide 98%
2-Methyl-N-tert-butylbenzamide
Formel:
C₁₂H₁₇NO Storage Temperature: Omgivande |
MDL Number:
MFCD00463017 CAS nummer: 104847-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...