Ethyl-2-amino-4-chlorobenzoate 98%
Leverantör: Apollo Scientific
OR907555-100G OR907555-25G OR907555-5G
APOSOR907555-100GEA 10175 SEK
APOSOR907555-100G APOSOR907555-25G APOSOR907555-5G
Ethyl-2-amino-4-chlorobenzoate 98%
Ethyl-2-amino-4-chlorobenzoate
Formel: C₉H₁₀ClNO₂
MW: 199,630 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09878359
CAS nummer: 60064-34-8