6-Propoxypyridazin-3-amine 95%
Leverantör: Apollo Scientific
OR906932-1G
OR906932-250MG
APOSOR906932-1GEA
540
SEK
APOSOR906932-1G
APOSOR906932-250MG
6-Propoxypyridazin-3-amine 95%
6-Propoxypyridazin-3-amine
Formel:
C₇H₁₁N₃O Storage Temperature: Omgivande |
MDL Number:
MFCD09835392 CAS nummer: 90008-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...