4,7-Dichlorothieno[2,3-d]pyridazine ≥98%
Leverantör: Apollo Scientific
OR905814-10G OR905814-1G OR905814-5G
APOSOR905814-10GEA 11900 SEK
APOSOR905814-10G APOSOR905814-1G APOSOR905814-5G
4,7-Dichlorothieno[2,3-d]pyridazine ≥98%
4,7-Dichlorothieno[2,3-d]pyridazine
Formel: C₆H₂Cl₂N₂S
MW: 205,0645 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09965522
CAS nummer: 699-89-8