5-Bromo-N-methylpyrimidin-4-amine 98%
Leverantör: Apollo Scientific
OR905716-1G OR905716-250MG
APOSOR905716-1GEA 6350 SEK
APOSOR905716-1G APOSOR905716-250MG
5-Bromo-N-methylpyrimidin-4-amine 98%
5-Bromo-N-methylpyrimidin-4-amine
Formel: C₅H₆BrN₃
MW: 188,03 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03861147
CAS nummer: 56181-38-5