2-(Aminomethyl)benzonitrile hydrochloride 96%
Leverantör: Apollo Scientific
OR905627-1G OR905627-250MG OR905627-5G
APOSOR905627-1GEA 1550 SEK
APOSOR905627-1G APOSOR905627-250MG APOSOR905627-5G
2-(Aminomethyl)benzonitrile hydrochloride 96%
2-(Aminomethyl)benzonitrile hydrochloride
Formel: C₈H₉ClN₂
MW: 168,6200 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD06797973
CAS nummer: 1134529-25-1