trans-1,4-Cyclohexanedimethanol 97%
Leverantör: Apollo Scientific
Synonyms: trans-1,4-Bis(hydroxymethyl)cyclohexane
OR904832-1G OR904832-25G OR904832-5G
APOSOR904832-1GEA 600 SEK
APOSOR904832-1G APOSOR904832-25G APOSOR904832-5G
trans-1,4-Cyclohexanedimethanol 97%
trans-1,4-Cyclohexanedimethanol
Formel: C₈H₁₆O₂
MW: 144,21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00066360
CAS nummer: 3236-48-4