1,3-Dimethyl-1H-pyrazol-4-carbaldehyde 98%
Leverantör: Apollo Scientific
Synonyms: 1,3-Dimethyl-1H-pyrazole-4-carbaldehyde
OR904769-1G OR904769-25G OR904769-5G
APOSOR904769-1GEA 1475 SEK
APOSOR904769-1G APOSOR904769-25G APOSOR904769-5G
1,3-Dimethyl-1H-pyrazol-4-carbaldehyde 98%
1,3-Dimethyl-1H-pyrazol-4-carbaldehyde
Formel: C₆H₈N₂O
MW: 124,1405 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00159635
CAS nummer: 25016-12-0