Methyl tetrahydro-2H-pyran-3-carboxylate 98%
Leverantör: Apollo Scientific
OR904288-1G OR904288-250MG
APOSOR904288-1GEA 5125 SEK
APOSOR904288-1G APOSOR904288-250MG
Methyl tetrahydro-2H-pyran-3-carboxylate 98%
Methyl tetrahydro-2H-pyran-3-carboxylate
Formel: C₇H₁₂O₃
MW: 144.1684 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09839011
CAS nummer: 18729-20-9