4-Nitroisoxazole 96%

Leverantör: Apollo Scientific

OR904227-1G OR904227-5G
APOSOR904227-1GEA 2400 SEK
APOSOR904227-1G APOSOR904227-5G
4-Nitroisoxazole 96%
4-Nitroisoxazole
Formel: C₃H₂N₂O₃
Storage Temperature: Kylskåp
MDL Number: MFCD06638027
CAS nummer: 1121-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR