3,5,6-Trichloropyridin-2(1H)-one 95%

Leverantör: Apollo Scientific

Synonyms: 3,5,6-Trichloro-2-pyridinol

OR903819-10G OR903819-1G OR903819-5G
APOSOR903819-10GEA 1175 SEK
APOSOR903819-10G APOSOR903819-1G APOSOR903819-5G
3,5,6-Trichloropyridin-2(1H)-one 95%
3,5,6-Trichloropyridin-2(1H)-one
Formel: C5H2Cl3NO
MW: 198,4345 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00130269
CAS nummer: 6515-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR