2-Amino-6-ethoxybenzothiazole 98%
Leverantör: Apollo Scientific
|
Varning
|
OR903400-100G
OR903400-25G
APOSOR903400-100GEA
720
SEK
APOSOR903400-100G
APOSOR903400-25G
2-Amino-6-ethoxybenzothiazole 98%
2-Amino-6-ethoxybenzothiazole
Formel:
C₉H₁₀N₂OS Storage Temperature: Omgivande |
MDL Number:
MFCD00005788 CAS nummer: 94-45-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...