3H-Pyrrolo[3,2-d]pyrimidin-4(5H)-one 95%
Leverantör: Apollo Scientific
Synonyms: 1,5-Dihydro-4h-pyrrolo[3,2-d]pyrimidin-4-one
OR903256-1G OR903256-250MG
APOSOR903256-1GEA 1200 SEK
APOSOR903256-1G APOSOR903256-250MG
3H-Pyrrolo[3,2-d]pyrimidin-4(5H)-one 95%
3H-Pyrrolo[3,2-d]pyrimidin-4(5H)-one
Formel: C₆H₅N₃O
MW: 135,13 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09832196
CAS nummer: 5655-01-6