5-Cyclopropyl-1H-pyrazole-3-carboxylic acid 96%
Leverantör: Apollo Scientific
OR903172-10G OR903172-1G OR903172-5G
APOSOR903172-10GEA 4275 SEK
APOSOR903172-10G APOSOR903172-1G APOSOR903172-5G
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid 96%
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid
Formel: C₇H₈N₂O₂
MW: 152,15 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04967194
CAS nummer: 401629-04-7