2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine 98%
Leverantör: Apollo Scientific
Synonyms: 2-[4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl]-5-(n-octyloxy)phenol, 2-(4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl)-5-(octyloxy)phenol
OR903058-25G OR903058-5G
APOSOR903058-25GEA 4725 SEK
APOSOR903058-25G APOSOR903058-5G
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine 98%
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine
Formel: C₃₃H₃₉N₃O₂
MW: 509,69 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09834146
CAS nummer: 2725-22-6