6-Amino-3-cyano-2-methylpyridine

Leverantör: Apollo Scientific

Synonyms: 6-Amino-2-methylnicotinonitrile

OR9028-25G OR9028-5G
APOSOR9028-25GEA 8325 SEK
APOSOR9028-25G APOSOR9028-5G
6-Amino-3-cyano-2-methylpyridine
6-Amino-3-cyano-2-methylpyridine
Formel: C₇H₇N₃
MW: 133,15 g/mol
Smältpunkt: 155…156 °C
Storage Temperature: Omgivande
MDL Number: MFCD05865116
CAS nummer: 183428-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR