7-Bromo-2-chlorothieno[3,2-d]pyrimidine ≥98%
Leverantör: Apollo Scientific
OR902544-100MG OR902544-1G OR902544-250MG
APOSOR902544-100MGEA 3525 SEK
APOSOR902544-100MG APOSOR902544-1G APOSOR902544-250MG
7-Bromo-2-chlorothieno[3,2-d]pyrimidine ≥98%
7-Bromo-2-chlorothieno[3,2-d]pyrimidine
Formel: C₆H₂BrClN₂S
MW: 249,5200 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD13193398
CAS nummer: 1152475-42-7