Methyl-3-hydroxyisoxazole-5-carboxylate 95%
Leverantör: Apollo Scientific
OR902507-1G
OR902507-5G
APOSOR902507-1GEA
0
SEK
APOSOR902507-1G
APOSOR902507-5G
Methyl-3-hydroxyisoxazole-5-carboxylate 95%
Methyl-3-hydroxyisoxazole-5-carboxylate
Formel:
C₅H₅NO₄ Storage Temperature: Omgivande |
MDL Number:
MFCD00062870 CAS nummer: 10068-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...