1-(Piperidin-4-yl)ethanone hydrochloride 96%
Leverantör: Apollo Scientific
OR902214-1G OR902214-5G
APOSOR902214-1GEA 720 SEK
APOSOR902214-1G APOSOR902214-5G
1-(Piperidin-4-yl)ethanone hydrochloride 96%
1-(Piperidin-4-yl)ethanone hydrochloride
Formel: C₇H₁₄ClNO
MW: 163.65 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00044810
CAS nummer: 89895-06-7