(N-Formyl-N'-(6-chloropyrid-2yl)hydrazine) 98%
Leverantör: Apollo Scientific
Synonyms: N'-(6-Chloropyridin-2-yl)formohydrazide
OR901597-1G OR901597-25G OR901597-5G
APOSOR901597-1GEA 1475 SEK
APOSOR901597-1G APOSOR901597-25G APOSOR901597-5G
(N-Formyl-N'-(6-chloropyrid-2yl)hydrazine) 98%
(N-Formyl-N'-(6-chloropyrid-2yl)hydrazine)
Formel: C₆H₆ClN₃O
Storage Temperature: Omgivande
MDL Number: MFCD11706996
CAS nummer: 66999-50-6