1-Benzylpyrrolidin-3-ylmethanol 97%
Leverantör: Apollo Scientific
Synonyms: 1-(Phenylmethyl)-3-pyrrolidinemethanol
OR901559-1G OR901559-25G OR901559-5G
APOSOR901559-1GEA 755 SEK
APOSOR901559-1G APOSOR901559-25G APOSOR901559-5G
1-Benzylpyrrolidin-3-ylmethanol 97%
1-Benzylpyrrolidin-3-ylmethanol
Formel: C₁₂H₁₇NO
MW: 191,28 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04038527
CAS nummer: 5731-17-9