4-Aminobenzyl alcohol 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol
OR9011-100G OR9011-25G OR9011-5G
APOSOR9011-100GEA 1275 SEK
APOSOR9011-100G APOSOR9011-25G APOSOR9011-5G
4-Aminobenzyl alcohol 98%
4-Aminobenzyl alcohol
Formel: C₇H₉NO
MW: 123.15 g/mol
Smältpunkt: 61…63 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00014782
CAS nummer: 623-04-1
EINECS: 210-767-5