4-Bromothiophene-2-methanol 98%
Leverantör: Apollo Scientific
Synonyms: 4-Bromo-2-thiophenemethanol, (4-Bromothiophen-2-yl)methanol, 4-Bromo-2-(hydroxymethyl)thiophene
OR900842-100G OR900842-25G OR900842-5G
APOSOR900842-100GEA 10175 SEK
APOSOR900842-100G APOSOR900842-25G APOSOR900842-5G
4-Bromothiophene-2-methanol 98%
4-Bromothiophene-2-methanol
Formel: C₅H₅BrOS
MW: 193,07 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04115392
CAS nummer: 79757-77-0