2-(4-Bromophenyl)propan-2-amine 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Bromo-alpha,alpha-dimethylbenzylamine
OR900669-10G OR900669-1G OR900669-5G
APOSOR900669-10GEA 12650 SEK
APOSOR900669-10G APOSOR900669-1G APOSOR900669-5G
2-(4-Bromophenyl)propan-2-amine 98%
2-(4-Bromophenyl)propan-2-amine
Formel: C₉H₁₂BrN
MW: 214,11 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD06797600
CAS nummer: 17797-12-5