(R)-(-)-Ethyl-3-piperidinecarboxylate
Leverantör: Apollo Scientific
Fara
Synonyms: Ethyl (R)-(-)-3-Piperidinecarboxylate, (R)-Ethyl piperidine-3-carboxylate
OR8969-100G OR8969-25G OR8969-5G
APOSOR8969-100GEA 2200 SEK
APOSOR8969-100G APOSOR8969-25G APOSOR8969-5G
(R)-(-)-Ethyl-3-piperidinecarboxylate
(R)-(-)-Ethyl-3-piperidinecarboxylate
Formel: C₈H₁₅NO₂
MW: 157,2102 g/mol
Densitet: 1,092 g/cm³ (25 °C)
Flampunkt: 104 °C
Storage Temperature: Kylskåp
MDL Number: MFCD03093637
CAS nummer: 25137-01-3