(R)-(+)-2-Pyrrolidinone-5-carboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid
OR8966-100G OR8966-25G OR8966-500G
APOSOR8966-100GEA 635 SEK
APOSOR8966-100G APOSOR8966-25G APOSOR8966-500G
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid
Formel: C₅H₇NO₃
MW: 129.12 g/mol
Smältpunkt: 155…156 °C
Storage Temperature: Omgivande
MDL Number: MFCD00066212
CAS nummer: 4042-36-8
EINECS: 223-735-0