Acetonylaceton 98%

Leverantör: Apollo Scientific
Varning

Synonyms: Hexane-2,5-dione, 2,5-Hexanedione

OR8860-100G OR8860-500G
APOSOR8860-100GEA 1125 SEK
APOSOR8860-100G APOSOR8860-500G
Acetonylaceton 98%
Acetonylaceton
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 185…192 °C
Smältpunkt: –6 to –5 °C
Densitet: 0,973 g/cm³ (25 °C)
Flampunkt: 79 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008792
CAS nummer: 110-13-4
EINECS: 203-738-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR