tert-Butyl-4-bromo-2-chlorobenzoate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: tert-Butyl 4-bromo-2-chlorobenzoate
OR8566-10G OR8566-1G OR8566-5G
APOSOR8566-10GEA 7800 SEK
APOSOR8566-10G APOSOR8566-1G APOSOR8566-5G
tert-Butyl-4-bromo-2-chlorobenzoate 98%
tert-Butyl-4-bromo-2-chlorobenzoate
Formel: C₁₁H₁₂BrClO₂
Storage Temperature: Omgivande
MDL Number: MFCD08703428
CAS nummer: 929000-18-0