tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
1-Boc-4-(4-bromophenyl)piperazine, 4-(4-Bromophenyl)piperazine-1-carboxylic acid tert-Butyl ester, 1-tert-Butoxycarbonyl-4-(4-bromophenyl)piperazine
OR8542-1G
OR8542-5G
APOSOR8542-1GEA
720
SEK
APOSOR8542-1G
APOSOR8542-5G
tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate 98%
tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate
Formel:
C₁₅H₂₁BrN₂O₂ MW: 341,25 g/mol Smältpunkt: 146…147 °C Storage Temperature: Omgivande |
MDL Number:
MFCD07371649 CAS nummer: 352437-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...